MilliporeSigma
  • Characterization of a PGA-based scaffold for use in a tissue-engineered neo-urinary conduit.
Characterization of a PGA-based scaffold for use in a tissue-engineered neo-urinary conduit.
Methods in molecular biology (Clifton, N.J.) (2013-03-16)
Toyin A Knight, Richard G Payne
ABSTRACT
A tissue-engineered product needs to be properly characterized in order to be used in vivo. Many methods can be used to characterize a scaffold during creation of a tissue-engineered product. This chapter looks at the mechanical (tensile testing) and biological characterization (cell viability and proliferation) of a polyglycolic acid-based scaffold used to tissue engineer a Neo-Urinary Conduit™. Such methods are more broadly applicable to characterization of other neo-organ product candidates.