MilliporeSigma
  • Functionalization of biodegradable magnesium alloy implants with alkylphosphonate self-assembled films.
Functionalization of biodegradable magnesium alloy implants with alkylphosphonate self-assembled films.
Materials science & engineering. C, Materials for biological applications (2013-03-19)
Z Grubač, M Metikoš-Huković, R Babić, I Škugor Rončević, M Petravić, R Peter
ABSTRACT
Mg and Mg-alloys are promising materials for biodegradable implants. In order to slowdown the Mg-alloy (AZ91D) degradation and enhance its biocompatibility, the alloy surface was modified with alkylphosphonate self-assembling films. The binding configuration and the structural organization of alkylphosphonate monolayers on the Mg-alloy surface were investigated using contact angle measurements, FTIR, and XPS. Combination of FTIR and XPS data indicated the presence of several different bonding modes (mono-, di-, and tri dentate) of phosphonate head groups with the alloy surface. The existence of well organized and ordered self-assembled alkylphosphonate monolayers with good barrier protecting properties in a physiological solution is a key step in the development of biocompatible Mg-alloy implants.
Sigma-Aldrich
Phosphorous acid solution, ≥50%
Sigma-Aldrich
Phosphorous acid, 99%