MilliporeSigma
  • Protein phosphatase 2A and DNA-dependent protein kinase are involved in mediating rapamycin-induced Akt phosphorylation.
Protein phosphatase 2A and DNA-dependent protein kinase are involved in mediating rapamycin-induced Akt phosphorylation.
The Journal of biological chemistry (2013-03-29)
Yikun Li, Xuerong Wang, Ping Yue, Hui Tao, Suresh S Ramalingam, Taofeek K Owonikoko, Xingming Deng, Ya Wang, Haian Fu, Fadlo R Khuri, Shi-Yong Sun
ABSTRACT
The mechanisms underlying rapamycin-induced Akt phosphorylation have not been fully elucidated. Inhibition of PP2A or DNA-PK attenuates or abrogates rapamycin-induced Akt phosphorylation and co-inhibition of mTOR and DNA-PK enhances anticancer activity. PP2A-dependent and DNA-PK-mediated mechanism is involved in rapamycin-induced Akt phosphorylation. A previously unknown mechanism underlying rapamycin-induced Akt phosphorylation and a novel strategy to enhance mTOR-targeted cancer therapy may be suggested. Inhibition of mammalian target of rapamycin complex 1 (mTORC1), for example with rapamycin, increases Akt phosphorylation while inhibiting mTORC1 signaling. However, the underlying mechanisms have not been fully elucidated. The current study has uncovered a previously unknown mechanism underlying rapamycin-induced Akt phosphorylation involving protein phosphatase 2A (PP2A)-dependent DNA protein kinase (DNA-PK) activation. In several cancer cell lines, inhibition of PP2A with okadaic acid, fostriecin, small T antigen, or PP2A knockdown abrogated rapamycin-induced Akt phosphorylation, and rapamycin increased PP2A activity. Chemical inhibition of DNA-PK, knockdown or deficiency of DNA-PK catalytic subunit (DNA-PKcs), or knock-out of the DNA-PK component Ku86 inhibited rapamycin-induced Akt phosphorylation. Exposure of cancer cells to rapamycin increased DNA-PK activity, and gene silencing-mediated PP2A inhibition attenuated rapamycin-induced DNA-PK activity. Collectively these results suggest that rapamycin induces PP2A-dependent and DNA-PK-mediated Akt phosphorylation. Accordingly, simultaneous inhibition of mTOR and DNA-PK did not stimulate Akt activity and synergistically inhibited the growth of cancer cells both in vitro and in vivo. Thus, our findings also suggest a novel strategy to enhance mTOR-targeted cancer therapy by co-targeting DNA-PK.
Sigma-Aldrich
Okadaic acid from Prorocentrum concavum, 92-100% (HPLC)