MilliporeSigma
  • A web-based route planning tool to reduce cyclists' exposures to traffic pollution: a case study in Montreal, Canada.
A web-based route planning tool to reduce cyclists' exposures to traffic pollution: a case study in Montreal, Canada.
Environmental research (2013-04-09)
Marianne Hatzopoulou, Scott Weichenthal, Guillaume Barreau, Mark Goldberg, William Farrell, Dan Crouse, Nancy Ross
ABSTRACT
We developed a web-based route planning tool for cyclists in Montreal, Canada, using spatial monitoring data for ambient nitrogen dioxide (NO2). With this tool, we estimated exposures to NO2 along shortest routes and lower exposure alternatives using origin-destination survey data. On average, exposures were estimated to be lower by 0.76 ppb (95% CI: 0.72, 0.80) relative to the shortest route, with decreases of up to 6.1 ppb for a single trip. Cumulative exposure levels (ppb km) decreased by approximately 4%. In general, the benefits of decreased exposure could be achieved with little increase (less than 1 km) in the overall route length.