MilliporeSigma
  • Saquinavir-NO inhibits S6 kinase activity, impairs secretion of the encephalytogenic cytokines interleukin-17 and interferon-gamma and ameliorates experimental autoimmune encephalomyelitis.
Saquinavir-NO inhibits S6 kinase activity, impairs secretion of the encephalytogenic cytokines interleukin-17 and interferon-gamma and ameliorates experimental autoimmune encephalomyelitis.
Journal of neuroimmunology (2013-04-23)
Filip Petković, Jana Blaževski, Miljana Momčilović, Gordana Timotijević, Mai-Britt Zocca, Sanja Mijatović, Danijela Maksimović-Ivanić, Katia Mangano, Paolo Fagone, Stanislava Stošić-Grujičić, Ferdinando Nicoletti, Djordje Miljković
ABSTRACT
NO-hybridization of the HIV protease inhibitor Saquinavir generates a new chemical entity named Saq-NO, that retains the anti-viral activity and exerts lower toxicity. We show that Saq-NO inhibited the generation of various cytokines in ConA-stimulated unfractionated murine spleen cells and rat lymph nodes stimulated with ConA as well as in purified CD4(+) T cells in vitro and reduced the circulating levels of cytokines in mice challenged with anti-CD3 antibody. Furthermore, Saq-NO reduced IL-17 and IFN-γ production in myelin basic protein (MBP)-specific cells isolated from rats immunized with MBP. These findings translated well into the in vivo setting as Saq-NO ameliorated the course of the disease in two preclinical models of multiple sclerosis. Our results demonstrate that Saq-NO exerts immunomodulatory effects that warrant studies on its application in autoimmune diseases.
Sigma-Aldrich
Saquinavir mesylate, ≥98% (HPLC), powder