MilliporeSigma
  • Bone morphogenetic protein-7 inhibits silica-induced pulmonary fibrosis in rats.
Bone morphogenetic protein-7 inhibits silica-induced pulmonary fibrosis in rats.
Toxicology letters (2013-05-04)
Gengxia Yang, Zhonghui Zhu, Yan Wang, Ai Gao, Piye Niu, Lin Tian
ABSTRACT
Bone morphogenetic protein-7 (BMP-7) has been shown to inhibit liver and renal fibrosis in in vivo and vitro studies. There is no study to investigate BMP-7's role in the development of pulmonary fibrosis induced by silica. In the current study, we used the rat model to explore the potential antifibrotic role of BMP-7 and its underlying mechanism in silica-induced pulmonary fibrosis. Sixty Wistar rats were randomly assigned into three groups. Control group received saline, silica group received silica and BMP-7 treated group received silica and BMP-7. BMP-7 was administered to silica-treated rats intraperitoneally at a dose of 300μg/kg/injection from day 8 to day 30 every other day. After the animals were sacrificed on day 15 and 30, hydroxyproline levels, the protein expressions of BMP/Smad and TGF-β/Smad signaling, and histopathology in lung tissues were analyzed. The hydroxyproline contents in BMP-7 treated groups were significantly lower than the silica groups (P<0.05). Histopathological results showed BMP-7 could reduce the progression of silica induced fibrosis. Furthermore, the expression of p-Smad1/5/8, a marker of BMP/Smad signaling, was significantly up-regulated in BMP-7 treated groups (P<0.05) compared with the silica groups. On the contrary, the expression of p-Smad2/3, a marker for TGF-β/Smad signaling, reduced significantly in BMP-7-treated groups compared with silica groups (P<0.05). In conclusion, the pulmonary fibrosis induced by silica in rats was significantly reduced with the therapeutic treatment of BMP-7. The antifibrotic effect of BMP-7 could be related to the activation of BMP/Smad signaling and inhibition of TGF-β/Smad pathways.
Sigma-Aldrich
trans-4-Hydroxy-L-proline, ≥99%
Sigma-Aldrich
trans-4-Hydroxy-L-proline, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
trans-4-Hydroxy-L-proline, BioReagent, suitable for cell culture, ≥98.5%
Supelco
trans-4-Hydroxy-L-proline, analytical standard