MilliporeSigma
Interaction of terminal transferase with single-stranded DNA.
The Journal of biological chemistry (1987-07-15)
D J Robbins, M D Barkley, M S Coleman
ABSTRACT
A 58-kDa monomer of terminal transferase was isolated from calf thymus using a monoclonal antibody affinity column. The enzymatic activity was comparable to that of the 44-kDa alpha beta dimer isolated by conventional methods. Binding of the two enzyme forms to single-stranded DNA was monitored by fluorescence. The site size of both forms was approximately 11 +/- 2 nucleotides. Binding of the 44-kDa alpha beta dimer to polydeoxyadenosine was examined under several conditions. The cooperativity parameter increased from about 90 in the presence of Mg2+ to 300-400 in the absence of Mg2+. The observed dissociation constant of 3-5 microM was essentially independent of salt concentration, whereas the intrinsic dissociation constant decreased about 5-fold in the presence of Mg2+. The binding parameters of the 58-kDa monomer were independent of buffer composition and were similar to those of the 44-kDa alpha beta dimer in the presence of Mg2+. These results indicate that the additional 14-kDa peptide sequences present in the high molecular mass monomer form are not part of the DNA-binding site of terminal transferase.