MilliporeSigma
  • Effect of sponge volume fraction on the performance of a novel fluidized bed bioreactor.
Effect of sponge volume fraction on the performance of a novel fluidized bed bioreactor.
Water science and technology : a journal of the International Association on Water Pollution Research (2013-06-12)
T T Nguyen, H H Ngo, W S Guo
ABSTRACT
A novel fluidized bed bioreactor (FBBR) was designed by integration of anaerobic granular activated carbon and aerobic sponge reactors. This FBBR was evaluated at different sponge volume fractions for treating a synthetic wastewater. Polyester urethane sponge with cube size of 1 × 1 × 1 cm and density of 28-30 kg/m(3) with 90 cells per 25 mm was used as biomass carrier. The results indicate that the FBBR could remove more than 93% of dissolved organic carbon (DOC). The highest nutrient removal efficiencies (58.2% PO4-P and 75.4% NH4-N) were achieved at 40% sponge volume fraction. The system could provide a good condition for biomass growth (e.g. 186.2 mg biomass/g sponge). No significant different performance in specific oxygen uptake rate was observed between 30, 40, and 50% sponge volume fractions.
Sigma-Aldrich
Phosphorus, red, ≥99.99% trace metals basis
Sigma-Aldrich
Phosphorus, red, ≥97.0%