MilliporeSigma
  • Opening the black box: revealing the molecular basis of thyroid hormone transport.
Opening the black box: revealing the molecular basis of thyroid hormone transport.
Endocrinology (2013-06-26)
Alexandro J Martagón, Kevin J Philips, Paul Webb
Sigma-Aldrich
L-Histidine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Histidine, ReagentPlus®, ≥99% (TLC)
Supelco
L-Histidine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Histidine, suitable for cell culture, meets EP, USP testing specifications, from non-animal source
SAFC
L-Histidine
Supelco
L-Histidine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland