MilliporeSigma
  • [Correlation between thermostability of the xylanase EvXyn11(TS) and its N-terminal disulfide bridge].
[Correlation between thermostability of the xylanase EvXyn11(TS) and its N-terminal disulfide bridge].
Wei sheng wu xue bao = Acta microbiologica Sinica (2013-07-19)
Rou Min, Jianfang Li, Shujuan Gao, Huimin Zhang, Jing Wu, Minchen Wu
ABSTRACT
To reveal the correlation between thermostability of xylanase EvXyn11(TS) and its N-terminal disulfide bridge, an EvXyn11(TS)-encoding gene (Syxyn11) was synthesized and subjected to site-directed mutagenesis. Multiple homology alignment of protein primary structures between the EvXyn11(TS) and several GH family 11 xylanases displayed that, in their N-termini, only EvXyn11(TS) contained a disulfide bridge (Cys5-Cys32), whose effect on the xylanase thermostability was predicted by molecular dynamics simulation. We constructed a gene Syxyn11(M), encoding the mutated xylanase (EvXyn11(M)) without N-terminal disulfide bridge. Then, Syxyn11 and Syxyn11(M) were expressed in Pichia pastoris GS115, and temperature and pH properties of the expressed enzymes were analyzed. The analytical results displayed that the temperature optimum of EvXyn11(M) was 70 degrees C, which was 15 degrees C lower than that of EvXyn11(TS). The half-life (t1/2(90)) of EvXyn11(TS) at 90 degrees C was 32 min, while the t1/2(70) of EvXyn11(M) at 70 degrees C was only 8.0 min. The important role of the N-terminal disulfide bridge on the thermostability of EvXyn11(TS) was first predicted by molecular dynamics simulation, and confirmed by site-directed mutagenesis. This work provided a novel strategy to improve thermostabilities of the mesophilic family 11 xylanases with high specific activities.
Sigma-Aldrich
Xylanase from Trichoderma viride, lyophilized powder, ≥100 units/mg protein