MilliporeSigma
  • Aminothiadiazole (NSC 4728) in patients with advanced carcinoma of the endometrium. A phase II study of the Gynecologic Oncology group.
Aminothiadiazole (NSC 4728) in patients with advanced carcinoma of the endometrium. A phase II study of the Gynecologic Oncology group.
American journal of clinical oncology (1990-02-01)
R F Asbury, J A Blessing, W P McGuire, P Hanjani, R Mortel
ABSTRACT
Twenty-one evaluable patients with advanced carcinoma of the endometrium were treated with aminothiadiazole at a dosage of 125 mg/m2 weekly. Twelve had received prior chemotherapy. Seven patients had stable disease; 14 had increasing disease. There were two life-threatening toxic episodes. Aminothiadiazole used in this dosage and schedule has negligible activity in previously treated patients with carcinoma of the endometrium.
Sigma-Aldrich
2-Amino-1,3,4-thiadiazole, 97%