MilliporeSigma
Fonofos toxicosis and milk residues in dairy cattle.
Veterinary and human toxicology (1985-08-01)
W O Cook, T L Carson
ABSTRACT
An organophosphate toxicosis due to fonofos was diagnosed in an Iowa Dairy herd. Fonofos was detected in the feed source and in the rumen contents, liver, and kidney tissue of one cow that died acutely. Bulk tank milk contained detectable levels of fonofos.