MilliporeSigma
  • Detection and separation of fenpropathrin, flucythrinate, fluvalinate, and PP 321 by thin-layer chromatography.
Detection and separation of fenpropathrin, flucythrinate, fluvalinate, and PP 321 by thin-layer chromatography.
Journal - Association of Official Analytical Chemists (1989-05-01)
R Khazanchi, S K Handa
ABSTRACT
Four synthetic pyrethroids having alpha-cyano ester groups, i.e., fenpropathrin, flucythrinate, fluvalinate, and PP 321, are separated by thin-layer chromatography and detected by a new set of chromogenic reagents. Synthetic pyrethroids containing the alpha-cyano group react with sodium hydroxide to liberate cyanide which forms pink spots with o-dinitrobenzene and p-nitrobenzaldehyde. The detection limit is 0.1 microgram and the method can be applied for identification and confirmation of these synthetic pyrethroids in vegetables.
Product Description
Supelco
Flucythrinate, PESTANAL®, analytical standard, mixture of stereo isomers