MilliporeSigma
  • Effects of high glucose and sorbitol pathway on lipid peroxidation of erythrocytes.
Effects of high glucose and sorbitol pathway on lipid peroxidation of erythrocytes.
Hormone and metabolic research = Hormon- und Stoffwechselforschung = Hormones et metabolisme (1989-05-01)
Y Nagasaka, S Fujii, T Kaneko
Sigma-Aldrich
1-Acetyl-2-phenylhydrazine, ≥98%