MilliporeSigma
  • A group-specific inhibitor of lysosomal cysteine proteinases selectively inhibits both proteolytic degradation and presentation of the antigen dinitrophenyl-poly-L-lysine by guinea pig accessory cells to T cells.
A group-specific inhibitor of lysosomal cysteine proteinases selectively inhibits both proteolytic degradation and presentation of the antigen dinitrophenyl-poly-L-lysine by guinea pig accessory cells to T cells.
Journal of immunology (Baltimore, Md. : 1950) (1986-01-01)
S Buus, O Werdelin
ABSTRACT
A limited intralysosomal proteolytic degradation is probably a key event in the accessory cell processing of large protein antigens before their presentation to T cells. With the aid of highly specific inhibitors of proteinases, we have examined the role of proteolysis in the presentation of antigens by guinea pig accessory cells. The proteinase inhibitor benzyloxycarbonyl-phenylalanylalanine-diazomethyl-ketone, which selectively inhibits cysteine proteinases, was used to block this set of enzymes in cultured cells. We demonstrate that the selective inhibition of the cysteine proteinases of antigen-presenting cells causes a profound inhibition of both the proteolytic degradation and the presentation of the synthetic antigen dinitrophenyl-poly-L-lysine. In contrast, the presentation of another synthetic antigen, the copolymer of L-glutamic acid and L-alanine, was enhanced by the same inhibitor. Another inhibitor, pepstatin A, which selectively blocks aspartic proteinases, did not block the presentation of dinitrophenyl-poly-L-lysine. The results identify cysteine proteinases, probably lysosomal, as one of the groups of enzymes involved in antigen processing.
Sigma-Aldrich
Z-L-Lys-ONp hydrochloride