MilliporeSigma
  • Efficacy of glycoprotein inhibitors alone and in combination with trifluridine in the treatment of murine herpetic keratitis.
Efficacy of glycoprotein inhibitors alone and in combination with trifluridine in the treatment of murine herpetic keratitis.
Current eye research (1986-02-01)
Y J Gordon, K P Cheng, T Araullo-Cruz, E Romanowski, B J Johnson, H A Blough
ABSTRACT
The present study examined the anti-herpetic effect of the glycoprotein inhibitors, hydroxynorvaline and 2-deoxyglucose, alone and in combination with trifluridine on murine ocular herpes. Following ocular inoculation with a large dose of HSV-1 RE strain (10(6) pfu), ICR mice were treated during the acute infection with different therapeutic regimens, and their efficacy was evaluated by ocular virus titers, clinical grading of blepharo-conjunctivitis and histological evaluation of stromal keratitis and iridocyclitis. The results following a large dose HSV-1 inoculum demonstrated that trifluridine was the best single therapeutic agent. Hydroxynorvaline and 2-deoxyglucose had no effect at all. Combination therapy of the glycoprotein inhibitors with trifluridine was no better than trifluridine alone. The mouse HSV-1 keratitis model proved to be an effective, economical alternative to the rabbit model for the evaluation of new antiviral agents.
Sigma-Aldrich
DL-3-Hydroxynorvaline, ≥98% (TLC)