MilliporeSigma
Allergic contact dermatitis from amcinonide.
Contact dermatitis (1985-04-01)
R Hayakawa, K Matsunaga, C Ukei, K Hosokawa
ABSTRACT
Allergic contact dermatitis from topical corticosteroids has been regarded as rare. However, it should be considered in long-standing, resistant or worsening eczema in spite of topical steroid therapy. Cases of allergic dermatitis from amcinonide have been reported previously. We report an additional case.