MilliporeSigma
Synthesis of c-2, 3, 17 and 19-oxygenated androgens.
Journal of steroid biochemistry (1988-03-01)
V C Njar, T Arunachalam, G Spiteller, E Caspi
ABSTRACT
The synthesis of C19-androgens oxygenated at carbons 2, 3, 17 and 19 and of their deuterium or deuterium-tritium labeled analogs is described.