MilliporeSigma
Glutathione oxidation during peroxidase catalysed drug metabolism.
Chemico-biological interactions (1987-01-01)
V V Subrahmanyam, L G McGirr, P J O'Brien
ABSTRACT
The peroxidase catalyzed oxidation of certain drugs in the presence of glutathione (GSH) resulted in extensive oxidation to oxidized glutathione (GSSG). Extensive oxygen uptake ensued and thiyl radicals could be trapped. Only catalytic amounts of drugs were required indicating a redox cycling mechanism. Active drugs included phenothiazines, aminopyrine, p-phenetidine, acetaminophen and 4-N,N-(CH3)2-aminophenol. Other drugs, including dopamine and alpha-methyl dopa, did not catalyse oxygen uptake, nor were GSSG or thiyl radicals formed. Instead, GSH was depleted by GSH conjugate formation. Drugs of the former group, e.g. acetaminophen, aminopyrine or N,N-(CH3)2-aniline have also been found by other investigators to form GSSG and hydrogen peroxide when added to hepatocytes or when perfused through an isolated liver. Although cytochrome P-450 normally catalyses a two-electron oxidation of drugs, serious consideration should be given for some one-electron oxidation resulting in radical formation, oxygen activation and GSSG formation.
Sigma-Aldrich
4-Ethoxyaniline, 98%