MilliporeSigma
  • Evaluation of flucythrinate- and fenvalerate-impregnated ear tags and permethrin ear tapes for fly (Diptera: Muscidae) control on beef and dairy cattle in northwest Florida.
Evaluation of flucythrinate- and fenvalerate-impregnated ear tags and permethrin ear tapes for fly (Diptera: Muscidae) control on beef and dairy cattle in northwest Florida.
Journal of economic entomology (1986-02-01)
J A Hogsette, J P Ruff
Product Description
Supelco
Flucythrinate, PESTANAL®, analytical standard, mixture of stereo isomers