MilliporeSigma
Methylation of thymine residues during oligonucleotide synthesis.
Nucleic acids research (1985-01-25)
X Gao, B L Gaffney, M Senior, R R Riddle, R A Jones
ABSTRACT
Thymine residues in an oligodeoxyribonucleotide are subject to methylation at N3 by the internucleotide methyl phosphotriester linkages. This alkylation occurs most rapidly in the presence of a strong base such as DBU, but also takes place, at a much slower rate, during oligonucleotide synthesis.
Sigma-Aldrich
Trimethyl phosphate, ≥99%
Sigma-Aldrich
Trimethyl phosphate, 97%