MilliporeSigma
  • 3',5'-Cyclic nucleotide phosphodiesterase activity of the sulphatase A of ox liver.
3',5'-Cyclic nucleotide phosphodiesterase activity of the sulphatase A of ox liver.
Biochimica et biophysica acta (1981-02-13)
T Uchida, F Egami, A B Roy
ABSTRACT
The sulphatase A (aryl-sulphate sulphohydrolase, EC 3.1.6.1) of ox liver hydrolyses adenosine 3',5'-monophosphate (cyclic AMP) to adenosine 5'-phosphate at an optimum pH of approx. 4.3, close that for the hydrolysis of cerebroside sulphate, a physiological substrate for sulphatase A. The Km is 11.6 mM for cyclic AMP. On polyacrylamide gel electrophoresis sulphatase A migrates as a single protein band which coincides with both the arylsulphatase and phosphodiesterase activities, suggesting that these are due to a single protein. Cyclic AMP competitively inhibits the arylsulphatase activity of sulphatase A, showing that both activities are associated with a single active site on the enzyme. sulphatase A also hydrolyses guanosine 3',5'-monophosphate, but not uridine 3',5'-monophosphate nor adenosine 2',3'-monophosphate.
Sigma-Aldrich
Adenosine 2′:3′-cyclic monophosphate sodium salt, ≥93%