MilliporeSigma
  • Direct homocysteine biosynthesis from O-succinylhomoserine in Escherichia coli: an alternate pathway that bypasses cystathionine.
Direct homocysteine biosynthesis from O-succinylhomoserine in Escherichia coli: an alternate pathway that bypasses cystathionine.
Journal of bacteriology (1983-01-01)
M Simon, J S Hong
ABSTRACT
Mutations were found which enable Escherichia coli K-12 to form homocysteine in the absence of cystathionase. The formation of homocysteine in the mutant strains required cystathionine gamma-synthetase, the metB gene product, but bypassed the normal intermediate cystathionine. It is concluded that cystathionine gamma-synthetase catalyzes the formation of homocysteine directly from O-succinylhomoserine and an as-yet-unidentified sulfur donor. The mutation apparently causes the formation of this sulfur donor and has been named metQ. The expression of the metQ gene is under catabolite repression.
Sigma-Aldrich
O-Succinyl-L-homoserine