MilliporeSigma
  • Analysis of ethchlorvynol (Placidyl): evaluation of a comparison performed in a clinical laboratory.
Analysis of ethchlorvynol (Placidyl): evaluation of a comparison performed in a clinical laboratory.
Journal of analytical toxicology (1984-11-01)
R R Bridges, T A Jennison
ABSTRACT
Common techniques for analyzing ethchlorvynol (Placidyl) by colorimetry and gas liquid chromatography are evaluated. Matrix effects are thoroughly examined to determine their contribution to the validity of results.
Sigma-Aldrich
Tungstic acid, ≥98.0% (calcined substance, T), powder
Sigma-Aldrich
Tungstic acid, 99%