MilliporeSigma
Dominant lethal effects of n-butyl glycidyl ether in mice.
Mutation research (1983-12-01)
E B Whorton, T G Pullin, A F Frost, A Onofre, M S Legator, D S Folse
ABSTRACT
Using the dominant lethal assay, the ability of n-butyl glycidyl ether to induce mutations in male mice was investigated. No significant dose-related changes either in pregnancy rates or in average number of implants per pregnant female were found. However, while the results were not altogether conclusive, there was evidence of a significant increase in fetal death rates by the end of the first week after the highest dosage was administered.
Sigma-Aldrich
Butyl glycidyl ether, 95%