MilliporeSigma
  • Evaluation of domifen bromide in the treatment of acute infectious oral diseases.
Evaluation of domifen bromide in the treatment of acute infectious oral diseases.
International journal of clinical pharmacology research (1983-01-01)
F Scaglione, G Scarpazza, E Marchi, G Biella, F Fraschini
ABSTRACT
By means of a double blind, placebo controlled trial, we have studied 29 patients (14 M and 15 F) affected by different acute infectious oral diseases. Our results suggest that domifen bromide may be useful in treating infectious oral diseases in combination with antibiotics, and in relieving pain and inflammation.
Sigma-Aldrich
Domiphen bromide, 97%
Pricing and availability is not currently available.