MilliporeSigma
  • Biosynthesis of radiolabeled verruculogen by Penicillium simplicissimum.
Biosynthesis of radiolabeled verruculogen by Penicillium simplicissimum.
Applied and environmental microbiology (1982-03-01)
J B Day, P G Mantle
ABSTRACT
In surface culture of Penicillium simplicissimum, verruculogen was shown to be biosynthesized from the intact carbon skeletons of tryptophan and proline, isoprenoid derivatives of mevalonic acid, and a methyl group donated by methionine. Selected radiolabeled precursors (1 mCi) pulse-fed at the optimum stage of fermentation yielded verruculogen (specific activity, 5.89 X 10(2) microCi mmol-1) labeled in the prolyl and isoprenyl regions of the molecule and suitable for metabolic studies.