MilliporeSigma
  • [Hygienic evaluation of the possible use of 1,3-dichloro-5,5-dimethylhydantoin for the purpose of drinking water decontamination].
[Hygienic evaluation of the possible use of 1,3-dichloro-5,5-dimethylhydantoin for the purpose of drinking water decontamination].
Gigiena i sanitariia (1982-03-01)
A A Semenova, A A Korolov
Sigma-Aldrich
1,3-Dichloro-5,5-dimethylhydantoin, available chlorine 68 %