MilliporeSigma
  • (+/-)-1-(Adamantan-2-yl)-2-propanamine and other amines derived from 2-adamantanone.
(+/-)-1-(Adamantan-2-yl)-2-propanamine and other amines derived from 2-adamantanone.
Il Farmaco; edizione scientifica (1980-05-01)
E Mariani, P Schenone, F Bondavalli, E Lampa, E Marmo
ABSTRACT
The synthesis of (+/-)-1-(adamantan-2-yl)-2-propanamine (VI), its N,N-dimethyl derivative (VII) and other amines (VIII), (IX) and (X), starting from 2-adamantanone, is described. Some of these amines showed antiparkinson activity in mice.
Sigma-Aldrich
2-Adamantanone, ReagentPlus®, 99%