MilliporeSigma
  • The influence of a heavy infection with sensitive and resistant strains of Ostertagia circumcincta and with Trichostrongylus colubriformis on the pharmacokinetics of febantel in lambs.
The influence of a heavy infection with sensitive and resistant strains of Ostertagia circumcincta and with Trichostrongylus colubriformis on the pharmacokinetics of febantel in lambs.
Journal of veterinary pharmacology and therapeutics (1995-06-01)
J Landuyt, M Debackere, J Vercruysse, Q McKellar
ABSTRACT
Plasma concentrations of febantel and its major metabolites, fenbendazole, oxfendazole and fenbendazole sulphone, were determined after oral administration of 7.5 mg/kg febantel in lambs before and 28 days after infection with 100,000 L3 larvae of a benzimidazole (BZ)-sensitive or BZ-resistant strain of Ostertagia circumcincta or with 75,000 L3 larvae of a BZ-sensitive Trichostrongylus colubriformis strain. The febantel concentrations were always low, and in only a few samples were higher than the limit of detection. A mean decrease in the area under the curve (AUC) for the three metabolites of 10.2%, 16.4% and 4.9% in lambs infected, respectively, with BZ-sensitive O. circumcincta, BZ-resistant O. circumcincta and T. colubriformis was observed. The Cmax for all the metabolites was higher in the BZ-sensitive O. circumcincta group than in the naive sheep, while the Tmax occurred earlier. The Cmax and the Tmax values for all the metabolites were lower in the BZ-resistant O. circumcincta group than in their own naive controls. In the T. colubriformis group the Cmax values of the metabolites were lower and the Tmax occurred much later.
Product Description
Supelco
Febantel, VETRANAL®, analytical standard