MilliporeSigma
Modulation of streptonigrin cytotoxicity by nitroxide SOD mimics.
Free radical biology & medicine (1994-11-01)
M C Krishna, R F Halevy, R Zhang, P L Gutierrez, A Samuni
ABSTRACT
Nitroxides are cell-permeable, stable radicals that react readily with paramagnetic species such as transition metals or short-lived free radicals, though not generally with diamagnetic molecules. Nitroxides can undergo one-electron selective redox reactions and thereby potentially modify the activity of cytotoxic drugs. Streptonigrin (SN) toxicity requires bioreduction to yield the semiquinone radical, and the toxicity is reportedly mediated by transition metals and oxygen-derived reactive species via redox-cycling of the semiquinone intermediate. The present study shows that (1) nitroxides protected isolated DNA and also aerated or hypoxic bacterial cells from SN toxicity; (2) H2O2 potentiated the hypoxic cytotoxicity of the drug but inhibited the damage to aerated cells; (3) pretreatment of cells with H2O2 conferred some protection, but not when the drug alone was preexposed to H2O2; and (4) desferrioxamine and 2,2-dipyridyl, though neither diethylenetriamino pentaacetate, exogenous catalase, or superoxide dismutase, decreased SN-induced cell killing. The mechanisms by which nitroxides protect from SN toxicity involve both a selective radical-radical reaction with SN semiquinone and the reoxidation of reduced cellular transition metal ions. On the other hand, H2O2 appears to exert two opposing effects: (1) facilitation of cell killing by the Fenton reaction and (2) lowering the cellular level of reducing equivalents, thus inhibiting the bioreductive activation of SN.
Sigma-Aldrich
4-Amino-TEMPO, free radical, 97%