MilliporeSigma
  • Catalytic properties of carbonyl reductase from rabbit liver for analogs of acetohexamide and 4-acetylpyridine.
Catalytic properties of carbonyl reductase from rabbit liver for analogs of acetohexamide and 4-acetylpyridine.
Biochemistry and molecular biology international (1994-08-01)
Y Imamura, T Koga, T Higuchi, M Otagiri, E Sugino, S Hibino, S Nagumo, H Akita
ABSTRACT
A correlation was observed between the values of specificity constant (kcat/Km) of carbonyl reductase from rabbit liver for acetohexamide analogs and their partition coefficients. This result indicates that the hydrophobicity in straight-chain alkyl groups of acetohexamide analogs plays an important role in the catalytic activity and substrate-binding capacity of the enzyme. Furthermore, the double logarithmic plots of kcat/Km values of the enzyme for 4-acetylpyridine analogs with a straight-chain alkyl group up to five carbon atoms against their partition coefficients gave a straight line. On the other hand, the plots for 4-acetylpyridine analogs with a straight-chain alkyl group over five carbon atoms and with a branched-chain alkyl group were away from the straight line. It is reasonable to postulate that a hydrophobic pocket is located in the substrate-binding domain of the enzyme.
Sigma-Aldrich
4-Acetylpyridine, 97%