MilliporeSigma
[Exacerbation of pustular psoriasis in mercury poisoning].
Der Hautarzt; Zeitschrift fur Dermatologie, Venerologie, und verwandte Gebiete (1994-10-01)
J Wehner-Caroli, C Scherwitz, F Schweinsberg, G Fierlbeck
ABSTRACT
A patient suffering from long-standing pustular psoriasis of the palms was treated for 3 weeks with a mercury-containing drug. Exacerbation into generalized pustular psoriasis developed. Mercury levels in blood and urine were increased. After withdrawal of the mercury preparation, therapy with DMPA (2,3-Dimercapto-1-propane-sulfonic acid), a mercury antidote, was initiated, together with short-term treatment with aromatic retinoids and PUVA. Within a few days mercury levels decreased significantly and the skin lesions practically disappeared.
Sigma-Aldrich
Mercury(II) iodide, ACS reagent, ≥99.0%
Sigma-Aldrich
Mercury(II) iodide red, puriss. p.a., ACS reagent, red, ≥99.0% (RT)