MilliporeSigma
  • [The immunochemical therapy of warts and growths due to the papillomavirus].
[The immunochemical therapy of warts and growths due to the papillomavirus].
Revue roumaine de virologie (Bucharest, Romania : 1990) (1993-01-01)
V Topciu, I Fufezan, R Mihăilescu, N Stanciu
ABSTRACT
A complex immuno-chemical treatment (immuno-stimulation, blockage of virus replication, interferon induction) was applied to 23 patients with warts and vegetations with various localisations. Results confirmed the treatment efficiency.
Sigma-Aldrich
Moroxydine hydrochloride, 99%