MilliporeSigma
  • Pyrrolizidine alkaloids from Senecio leucanthemifolius and Senecio rodriguezii.
Pyrrolizidine alkaloids from Senecio leucanthemifolius and Senecio rodriguezii.
Natural toxins (1993-01-01)
E Roeder, T Bourauel
ABSTRACT
Two toxic pyrrolizidine alkaloids were isolated from Senecio leucanthemifolius and six from Senecio rodriguezii. Their structures were determined by spectroscopic methods.