MilliporeSigma
  • [Evaluation of Ricide-P pesticide toxicity using the experimental model mother-fetus-newborn].
[Evaluation of Ricide-P pesticide toxicity using the experimental model mother-fetus-newborn].
Gigiena i sanitariia (1993-08-01)
M V Golubchikova, T V Belianina, M M Korshun, V I Kalinina
ABSTRACT
Necessity of the mother-fetus-newborn model usage in toxicological experiments for the xenobiotics environmental content standardization was shown in the toxicological study of Ricid-P pesticide.
Product Description
Supelco
Iprobenfos, PESTANAL®, analytical standard