MilliporeSigma
  • Enzymatic synthesis of a quorum-sensing autoinducer through use of defined substrates.
Enzymatic synthesis of a quorum-sensing autoinducer through use of defined substrates.
Science (New York, N.Y.) (1996-06-14)
M I Moré, L D Finger, J L Stryker, C Fuqua, A Eberhard, S C Winans
ABSTRACT
Many bacteria, including several pathogens of plants and humans, use a pheromone called an autoinducer to regulate gene expression in a cell density-dependent manner. Agrobacterium autoinducer [AAI, N-(3-oxo-octanoyl)-L-homoserine lactone] of A. tumefaciens is synthesized by the Tral protein, which is encoded by the tumor-inducing plasmid. Purified hexahistidinyl-Tral (H6-Tral) used S-adenosylmethionine to make the homoserine lactone moiety of AAI, but did not use related compounds. H6-Tral used 3-oxo-octanoyl-acyl carrier protein to make the 3-oxo-octanoyl moiety of AAI, but did not use 3-oxo-octanoyl-coenzyme A. These results demonstrate the enzymatic synthesis of an autoinducer through the use of purified substrates.
Sigma-Aldrich
N-(3-Oxooctanoyl)-L-homoserine lactone, ≥97% (HPLC), white, powder