MilliporeSigma
  • Determination of synthetic estrogens in illegal veterinary formulations by HPTLC and HPLC.
Determination of synthetic estrogens in illegal veterinary formulations by HPTLC and HPLC.
Journal of chromatographic science (1996-07-01)
E S Nascimento, M C Salvadori, L M Ribeiro-Neto
ABSTRACT
To determine the actual amount of diethylstilbestrol, hexestrol, and dienestrol in formulations such as pellets and oily injections that are illegally available on the Brazilian market, a simple methanol extraction is used for the analysis of the pellets and an ether extraction with Sephadex columns (for clean-up) is used for the oily injections. High-performance thin-layer chromatography is used for identification (as a qualitative and semiquantitative method), and high-performance liquid chromatography is used for quantitation. The results of the analysis show that all the formulations are not in accordance with the information listed on their labels.