MilliporeSigma
  • Two amine oxidases from Aspergillus niger AKU 3302 contain topa quinone as the cofactor: unusual cofactor link to the glutamyl residue occurs only at one of the enzymes.
Two amine oxidases from Aspergillus niger AKU 3302 contain topa quinone as the cofactor: unusual cofactor link to the glutamyl residue occurs only at one of the enzymes.
Biochimica et biophysica acta (1996-06-07)
I Frébort, P Pec, L Luhová, H Toyama, K Matsushita, S Hirota, T Kitagawa, T Ueno, Y Asano, Y Kato, O Adachi
ABSTRACT
Amine oxidases (EC 1.4.3.6) from Aspergillus niger, AO-I (2 x 75 kDa) and AO-II (80 kDa), were examined to determine the cofactor structure. Inactivated with p-nitrophenylhydrazine, they showed absorption and fluorescence spectra similar to those published for other copper amine oxidases and to topa hydantoin p-nitrophenylhydrazone. After digestion by thermolysin and pronase, cofactor peptides were purified by HPLC and sequenced. For thermolytic peptides, a typical topa consensus sequence, Asn-X-Glu-Tyr, was obtained for AO-II, although in case of AO-I it overlapped with Val-Val-Ile-Glu-Pro-Tyr-Gly. For pronase peptides of AO-I, only the latter sequence was obtained. NMR and mass spectroscopy confirmed the residue X as topa p-nitrophenylhydrazone in AO-II and revealed the presence of a residue Z attached to the Glu in the peptide Val-Val-Ile-Glu(Z)-Pro of AO-I. This residue was separated from the peptide by hydrolysis and identified as a product derived from topa quinone. The data, together with amino-acid sequence of AO-I, confer strong evidence for topa quinone as the cofactor, bound in the typical consensus sequence. Raman spectra of the p-nitrophenylhydrazone derivative of AO-I and its pronase peptide showed essentially the same peaks matching to a model compound for topa p-nitrophenylhydrazone. However, there may exist an unusual ester link between the topa-404 and Glu-145 in the native enzyme.