MilliporeSigma
Synthesis of oxido-bridged analogs of 18-hydroxyprogesterone.
Steroids (1996-06-01)
M O Doctorovich, A A Ghini, G Burton
ABSTRACT
18-Hydroxy-6,19-oxidoprogesterone and 18-hydroxy-11, 19-oxidoprogesterone were synthesized from readily available materials. The functionalization of C-18 was accomplished with phenyliodosodiacetate/iodine, whereas that of C-19 was carried out with the mercuric oxide/iodine system, both under irradiation with visible light. For 18-hydroxy-11,19-oxidoprogesterone, C-19 was functionalized before C-18, whereas the reverse order had to be used for the 6,19-oxido derivative.
Sigma-Aldrich
Mercury(II) oxide yellow, ACS reagent, ≥99.0%
Sigma-Aldrich
Mercury(II) oxide, 99.999% trace metals basis
Sigma-Aldrich
Mercury(II) oxide red, ACS reagent, ≥99.0%