MilliporeSigma
  • Pemphigoid and pemphigus foliaceus successfully treated with topical corticosteroids.
Pemphigoid and pemphigus foliaceus successfully treated with topical corticosteroids.
The Journal of dermatology (1996-10-01)
T Muramatsu, T Iida, T Shirai
ABSTRACT
Six patients with pemphigoid and three patients with pemphigus foliaceus were successfully treated with topical corticosteroids. This was especially effective in cases of pretibial localized pemphigoid and in mild cases of bullous pemphigoid and pemphigus foliaceus with negative or low-titer circulating autoantibodies.