MilliporeSigma
  • Migration studies from paper and board food packaging materials. 1. Compositional analysis.
Migration studies from paper and board food packaging materials. 1. Compositional analysis.
Food additives and contaminants (1997-01-01)
L Castle, C P Offen, M J Baxter, J Gilbert
ABSTRACT
A survey of 32 paper and board materials intended for food contact has been undertaken. Sample composition was analysed to identify chemicals with a potential to migrate to foods. Representative materials covering a range of uses such as aqueous and fatty foods, and use at ambient and elevated temperatures, were obtained from industry. The samples were examined in their unfinished state, even where the final form would have included lamination to plastics, printing, or other treatments. A comprehensive and systematic analytical approach was applied which included determination of volatiles by headspace GC-MS, extraction using water, ethanol and chloroform with subsequent analysis by GC-MS and HPLC, and multi-element screening by ICP-MS. The solvent extracts were also weighed to determine the total amount of material available for migration. A large number of volatile substances were identified with alkyl and aryl aldehydes predominating in the majority of samples. Substances identified in solvent extracts included 2,6-di-tert-butyl-4-hydroxytoluene (BHT), di-tert-butylphenol, benzophenone, 4,4'-bis(dimethyl amino) benzophenone (Michler's ketone), triphenyl methane, bicyclohexylphenylphenanthrene carboxylic acid (and its methyl ester) and abietic acid. Levels of these substances were however generally below 1 mg/kg paper. Of the inorganic elements, arsenic and mercury were not detected above the limits of determination of 1.8 and 0.4 mg/kg respectively in any of the 10 samples examined. Cadmium was detected in teabag tissue and in unbleached Kraft paper at 0.3 mg/kg. Chromium was present in seven samples at 1.1-7.8 mg/kg, whilst lead was present in nine of the 10 samples at levels ranging from 0.3 to 5.9 mg/kg.
Sigma-Aldrich
Michler′s ketone, 98%