MilliporeSigma
  • Genetic evidence for the existence of two quinone related inhibitor binding sites in NADH-CoQ reductase.
Genetic evidence for the existence of two quinone related inhibitor binding sites in NADH-CoQ reductase.
Biochimica et biophysica acta (1997-03-28)
E Darrouzet, A Dupuis
ABSTRACT
Using the NADH-CoQ reductase of Rhodobacter capsulatus as a model for the mitochondrial Complex I, we have for the first time isolated bacterial mutants resistant to piericidin-A, a classical inhibitor of the mitochondrial enzyme. Their sensitivity to other inhibitors directed towards the quinone binding domain of complex I gives direct genetic evidence for the existence of two inhibitor binding sites.