MilliporeSigma
  • Metabolic activation and formation of DNA adducts of hexestrol, a synthetic nonsteroidal carcinogenic estrogen.
Metabolic activation and formation of DNA adducts of hexestrol, a synthetic nonsteroidal carcinogenic estrogen.
Chemical research in toxicology (1998-06-20)
S T Jan, P D Devanesan, D E Stack, R Ramanathan, J Byun, M L Gross, E G Rogan, E L Cavalieri
ABSTRACT
Hexestrol (HES), a synthetic nonsteroidal estrogen, is carcinogenic in Syrian golden hamsters. The major metabolite of HES is its catechol, 3'-OH-HES, which can be metabolically converted to the electrophilic catechol quinone, HES-3',4'-Q, by peroxidases and cytochrome P450. Standard adducts were synthesized by reacting HES-3',4'-Q with dG and dA to produce the adducts 3'-OH-HES-6'(alpha, beta)-N7Gua and HES-3',4'-Q-6'-N6dA, respectively. When HES-3',4'-Q was reacted with calf thymus DNA, 3'-OH-HES-6'(alpha,beta)-N7Gua was identified by HPLC and tandem mass spectrometry as the depurinating adduct, with minor amounts of stable adducts. 3'-OH-HES was bound to DNA after activation by horseradish peroxidase, lactoperoxidase, or rat liver microsomes. The depurinating adduct 3'-OH-HES-6'(alpha, beta)-N7Gua was identified in these systems at levels of 65, 41, and 11 micromol/mol of DNA-P, respectively. Unidentified stable adducts were observed in much lower amounts and were quantified by the 32P-postlabeling method. Similarly to 3'-OH-HES, the catechol metabolites of the natural steroidal estrogens estrone (E1) and estradiol (E2), namely, 2-OHE1, 4-OHE1, 2-OHE2, and 4-OHE2, can be oxidized to their corresponding quinones by peroxidases and cytochrome P450. The quinones of the carcinogenic 4-OHE1 and 4-OHE2 have chemical and biochemical properties similar to those of HES-3',4'-Q. The results suggest that formation of HES-3',4'-Q may be a critical event in tumor initiation by HES and that HES is an excellent model compound to corroborate the hypothesis that estrogen-3,4-quinones are ultimate carcinogenic metabolites of the natural steroidal estrogens E1 and E2.