MilliporeSigma
  • Synthesis of 1-(beta-D-glycopyranosyl)-3-deazapyrimidines from 2-hydroxy and 2-mercaptopyridines.
Synthesis of 1-(beta-D-glycopyranosyl)-3-deazapyrimidines from 2-hydroxy and 2-mercaptopyridines.
Nucleosides & nucleotides (1998-07-22)
A M Attia, G H Elgemeie, I S Alnaimi
ABSTRACT
The synthesis of new 4- and 5-substituted-3-cyanopyridine nucleosides has been performed by reacting the silylated pyridines and penta-omicron-acetyl-alpha -D-glycopyranose in dichloroethane in the presence of SnCl4. The free nucleosides were tested for their potential activity against HIV and different types of tumor.
Sigma-Aldrich
2-Mercaptopyridine, ReagentPlus®, 99%