MilliporeSigma
  • GTH II but not GTH I induces final maturation and the development of maturational competence of oocytes of red seabream in vitro.
GTH II but not GTH I induces final maturation and the development of maturational competence of oocytes of red seabream in vitro.
General and comparative endocrinology (1998-09-28)
H Kagawa, H Tanaka, K Okuzawa, M Kobayashi
ABSTRACT
The effects of red seabream gonadotropins (PmGTH I and PmGTH II) on the induction in vitro of germinal vesicle breakdown (GVBD) and the development of maturational competence (responsiveness to maturation-inducing steroid) were examined in the oocytes of red seabream. PmGTH II was highly effective in inducing GVBD in both maturationally incompetent (45.6 +/- 3.2% GVBD at a concentration of 300 ng/ml) and competent oocytes (42.3 +/- 0.4% GVBD at a concentration of 300 ng/ml). 17,20beta-Dihydroxy-4-pregnen-3-one (DHP, 10 ng/ml) increased the frequency of GVBD induced by PmGTH II. PmGTH I (33, 100, 300, and 900 ng/ml) was unable to induce GVBD at any tested concentration in the presence or absence of DHP. Actinomycin D (1 microgram/ml) and cycloheximide (1 microgram/ml) totally inhibited the PmGTH II-induced GVBD in the presence and in the absence of DHP. Both PmGTH I and PmGTH II stimulated in vitro production of 11-ketotestosterone in sliced testes of red seabream in a similar potency. These results indicate that PmGTH II, but not PmGTH I, induces the final maturation of oocytes, as well as the development of the maturational competence of oocytes, in red seabream.
Sigma-Aldrich
17α,20β-Dihydroxy-4-pregnen-3-one, crystalline