MilliporeSigma
  • Effect of side chains including the N-methyl-tetrazole-thiol group of beta-lactam antibiotics on transport in cultured kidney epithelial cells LLC-PK1.
Effect of side chains including the N-methyl-tetrazole-thiol group of beta-lactam antibiotics on transport in cultured kidney epithelial cells LLC-PK1.
Biological & pharmaceutical bulletin (1998-11-20)
K Goto, M Oda, H Saitoh, M Nishida, M Takada
ABSTRACT
In this study, the transcellular transport characteristics of four beta-lactam antibiotics (cefotaxime, cefmenoxime, cefmetazole, and cefotiam) were investigated in a kidney epithelial cell line LLC-PK1, especially focusing on the effect of the N-methyl-tetrazole-thiol (NMTT) group attached to 7-amino-cephalosporanic acid. There were no directional differences between the apical-to-basolateral and basolateral-to-apical transport of cefotaxime, cefmenoxime, and cefmetazole, suggesting that the NMTT group does not influence the transcellular transport behaviors of beta-lactam antibiotics. In contrast, cefotiam transport across LLC-PK1 cell monolayers was 1.3-fold greater in the basolateral-to-apical direction than in the apical-to-basolateral direction. It is considered that the ionization of nitrogen in the N-dimethylaminoethyl group attached to NMTT is a factor in the secretory-oriented movement of cefotiam. The transcellular transport of cefotiam in both directions was significantly depressed at a low temperature (4 degrees C) and by 2,4-dinitrophenol. The basolateral-to-apical transport of cefotiam was also shown to be concentration-dependent. These results suggest that a specialized transport process might participate in the transcellular transport of cefotiam. The lipophilicities of these beta-lactam antibiotics were not correlated to the degree of transcellular transport, directly.