MilliporeSigma
  • A simple loop method for the automated preparation of (11C)raclopride from (11C)methyl triflate.
A simple loop method for the automated preparation of (11C)raclopride from (11C)methyl triflate.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine (2001-05-08)
R Iwata, C Pascali, A Bogni, Y Miyake, K Yanai, T Ido
ABSTRACT
A simple automated preparation of [11C]raclopride by reaction of [11C]methyl triflate with demethylraclopride triflate is described. The conventional bubbling of [11C]methyl triflate into the precursor solution was compared with two alternative methods which used a commercially available C18 cartridge (on-column method) or an empty PTFE tube (loop method) as support for the precursor solution. The influence of several solvents was assessed for all three methods. The on-column method showed excellent trapping efficiencies of [11C]methyl triflate but gave the lowest radiochemical yields. The loop method proved to be a simplified alternative to the bubbling method, giving comparable radiochemical yields with less precursor and offering an easy way to transfer the reaction mixture into an HPLC column. By the simple-loop method [11C]raclopride could be prepared in over 40% radiochemical yields (decay-corrected and based on [11C]methyl triflate).
Sigma-Aldrich
Methyl trifluoromethanesulfonate, ≥98%