MilliporeSigma
  • [Study on the injury of liver induced by terephthalic acid ethylene glycol and/or dowtherm A in rats].
[Study on the injury of liver induced by terephthalic acid ethylene glycol and/or dowtherm A in rats].
Wei sheng yan jiu = Journal of hygiene research (2003-02-04)
Hongwei Yao, Xinru Wang, Xikun Xu, Aimin Shi
ABSTRACT
The joint injury actions and mechanisms of terephthalic acid (TPA), ethylene glycol (EG) and/or dowtherm A (DOW) on liver in rats were investigated. A subchronic toxicity study was designed by a 2(3) factorial method. Some enzymes, biochemical and morphologic indices reflecting the injury of liver were studied. The results showed that serum ALT and serum total bile acid (TBA) of rats in the combined intoxication groups were significantly higher than those in the groups with single toxic agent and control group. The results of factorial analysis showed that the joint action induced by TPA, EG and/or DOW were characterized as additive (TPA + EG), synergistic (EG + DOW), synergistic (TPA + DOW) and additive(TPA + EG + DOW) actions. The deduction was identified by morphologic observations.
Sigma-Aldrich
Dowtherm® A, eutectic mixture of 26.5% diphenyl + 73.5% diphenyl oxide