MilliporeSigma
  • Elucidation of the extraordinary 4-membered pyrrole ring-contracted azeteoporphyrinoid as an intermediate in chlorin oxidation.
Elucidation of the extraordinary 4-membered pyrrole ring-contracted azeteoporphyrinoid as an intermediate in chlorin oxidation.
Chemical communications (Cambridge, England) (2006-12-01)
Tillmann Köpke, Maren Pink, Jeffrey M Zaleski
ABSTRACT
Reaction of 2,3-dioxochlorins with benzeneselenic anhydride (BSA) results in the formation of unusual ring-contracted azetine derivatives that further react with BSA to afford porpholactones.